Learn More
Waters Corp Waters Oasis WCX Online Cartridge, 30 μm, 10 mm, 96/pk
Catalog No. 50466268 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The cartridges are designed for rugged, reproducible, and ultra-fast on-line solid phase extraction (SPE). They contain 2.5 mg of a polymeric reversed-phase, weak cation exchange mixed-mode sorbent, optimized for selective sample preparation of strong bases and quaternary amines.
  • Rugged, reproducible, and ultra-fast on-line SPE.
  • Contains 2.5 mg of WCX polymeric mixed-mode sorbent.
  • Optimized for selective sample preparation of strong bases and quaternary amines.
  • Each 800-series tray holds 96 cartridges.
  • Directly loadable into automatic cartridge exchange (ACE) or ACE feeder units.
  • Water-wettable sorbent for high retention and excellent recoveries.
  • Syringe-barrel-type for use with vacuum manifolds and automated SPE instruments.
  • Ultra-clean to prevent contamination.
  • Manufactured in a cGMP, ISO 9001 certified plant.
  • Wide operating pH range of 0 to 14.
Catalog No. 50-466-268 Supplier Waters Corp Supplier No. 186005784
The cartridges are designed for rugged, reproducible, and ultra-fast on-line solid phase extraction (SPE). They contain 2.5 mg of a polymeric reversed-phase, weak cation exchange mixed-mode sorbent, optimized for selective sample preparation of strong bases and quaternary amines.
  • Rugged, reproducible, and ultra-fast on-line SPE.
  • Contains 2.5 mg of WCX polymeric mixed-mode sorbent.
  • Optimized for selective sample preparation of strong bases and quaternary amines.
  • Each 800-series tray holds 96 cartridges.
  • Directly loadable into automatic cartridge exchange (ACE) or ACE feeder units.
  • Water-wettable sorbent for high retention and excellent recoveries.
  • Syringe-barrel-type for use with vacuum manifolds and automated SPE instruments.
  • Ultra-clean to prevent contamination.
  • Manufactured in a cGMP, ISO 9001 certified plant.
  • Wide operating pH range of 0 to 14.