Learn More
Waters Corp Waters Online Column, WCX, 30 μm, 80 Å, 2.1 mm x 20 mm
Catalog No. 50781162 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The WCX on-line column is engineered for rugged, reproducible, and ultra-fast Solid Phase Extraction (SPE). It uses a polymeric reversed-phase, weak ion exchange mixed-mode sorbent, optimized for selective sample preparation of strong basic compounds and quaternary amines. It is designed for use with narrow-bore analytical columns and traditional HPLC column fittings.
  • Rugged, reproducible, and ultra-fast on-line SPE.
  • Polymeric reversed-phase, weak ion exchange mixed-mode sorbent for highly selective sample preparation.
  • Broad operational pH range of 0 to 14.
  • Water-wettable sorbent maintains capacity even if it runs dry.
Catalog No. 50-781-162 Supplier Waters Corp Supplier No. 186002505
The WCX on-line column is engineered for rugged, reproducible, and ultra-fast Solid Phase Extraction (SPE). It uses a polymeric reversed-phase, weak ion exchange mixed-mode sorbent, optimized for selective sample preparation of strong basic compounds and quaternary amines. It is designed for use with narrow-bore analytical columns and traditional HPLC column fittings.
  • Rugged, reproducible, and ultra-fast on-line SPE.
  • Polymeric reversed-phase, weak ion exchange mixed-mode sorbent for highly selective sample preparation.
  • Broad operational pH range of 0 to 14.
  • Water-wettable sorbent maintains capacity even if it runs dry.