Learn More
Waters Corp Waters Corporation Oasis WCX 96-well Plate, 10 mg Sorbent per Well, 30 μm, 1/pk
Catalog No. 50781158 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Oasis WCX 96-well plate features a polymeric reversed-phase, weak ion exchange mixed-mode sorbent, optimized for highly selective sample preparation of strong basic compounds and quaternary amines. Its water-wettability ensures consistent results, compatible with robotic systems.
  • Polymeric reversed-phase, weak ion exchange mixed-mode sorbent.
  • Highly selective for strong basic compounds and quaternary amines.
  • Water-wettability ensures consistent results even if wells dry out.
  • Compatible with robotic liquid handling systems and various manifold configurations.
  • Stable from pH 0 to 14, allowing stronger washes and cleaner extracts.
  • Optimized design for faster flow rates with viscous samples.
  • Ensures high batch-to-batch and lot-to-lot reproducibility.
Catalog No. 50-781-158 Supplier Waters Corp Supplier No. 186002501
This Oasis WCX 96-well plate features a polymeric reversed-phase, weak ion exchange mixed-mode sorbent, optimized for highly selective sample preparation of strong basic compounds and quaternary amines. Its water-wettability ensures consistent results, compatible with robotic systems.
  • Polymeric reversed-phase, weak ion exchange mixed-mode sorbent.
  • Highly selective for strong basic compounds and quaternary amines.
  • Water-wettability ensures consistent results even if wells dry out.
  • Compatible with robotic liquid handling systems and various manifold configurations.
  • Stable from pH 0 to 14, allowing stronger washes and cleaner extracts.
  • Optimized design for faster flow rates with viscous samples.
  • Ensures high batch-to-batch and lot-to-lot reproducibility.