Learn More
Waters Corp Waters Corporation 96-well Plate, Oasis WCX, 30 mg Sorbent per Well, 30 μm
Catalog No. 50781160 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis WCX 96-well plate utilizes a polymeric reversed-phase, weak ion exchange mixed-mode sorbent, ideal for selective sample preparation of strong basic compounds and quaternary amines. Its water-wettability prevents issues from drying wells, while polypropylene construction ensures chemical stability. It is compatible with various manifold configurations and robotic systems.
  • Polymeric reversed-phase, weak ion exchange mixed-mode sorbent.
  • Optimized for selective sample preparation of strong basic compounds and quaternary amines.
  • Unique balance of hydrophobicity and water-wettability.
  • Compatible with many manifold configurations and most robotic liquid handling systems.
  • Constructed from polypropylene for chemical stability and inertness.
  • Offers broad operational pH range (0 to 14 pH).
  • Ensures exceptional product quality and reproducibility.
Catalog No. 50-781-160 Supplier Waters Corp Supplier No. 186002503
The Oasis WCX 96-well plate utilizes a polymeric reversed-phase, weak ion exchange mixed-mode sorbent, ideal for selective sample preparation of strong basic compounds and quaternary amines. Its water-wettability prevents issues from drying wells, while polypropylene construction ensures chemical stability. It is compatible with various manifold configurations and robotic systems.
  • Polymeric reversed-phase, weak ion exchange mixed-mode sorbent.
  • Optimized for selective sample preparation of strong basic compounds and quaternary amines.
  • Unique balance of hydrophobicity and water-wettability.
  • Compatible with many manifold configurations and most robotic liquid handling systems.
  • Constructed from polypropylene for chemical stability and inertness.
  • Offers broad operational pH range (0 to 14 pH).
  • Ensures exceptional product quality and reproducibility.