Learn More
Waters Corp Waters μElution Plate, WCX, 96-well, 2 mg/well, 30 μm
Catalog No. 50781156 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The patented 96-well Oasis WCX μElution plate is designed for SPE cleanup and analyte enrichment of sample volumes ranging from 10 μL to 375 μL.
  • Water-wettable polymeric sorbent stable from pH 0 to 14.
  • Exhibits excellent retention and capacity for a wider polarity spectrum of analytes.
Catalog No. 50-781-156 Supplier Waters Corp Supplier No. 186002499
The patented 96-well Oasis WCX μElution plate is designed for SPE cleanup and analyte enrichment of sample volumes ranging from 10 μL to 375 μL.
  • Water-wettable polymeric sorbent stable from pH 0 to 14.
  • Exhibits excellent retention and capacity for a wider polarity spectrum of analytes.