Learn More
Waters Corp Waters Vac Cartridge, WAX, 1 cc, 30 mg, 30 μm
Catalog No. 50819118 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 1 cc Oasis WAX cartridges contain the Oasis WAX sorbent, a polymeric reversed-phase, weak anion exchange mixed-mode sorbent optimized for fast, simple, and highly selective sample preparation of strong acidic compounds. The Oasis WAX sorbent is water wettable, maintaining its capability for higher retention and excellent recoveries even if the sorbent runs dry, eliminating the need for extraordinary steps to prevent drying out during critical steps prior to sample loading.
  • Polymeric reversed-phase, weak anion exchange mixed-mode sorbent for strong acidic compounds
  • Water wettable to maintain retention and recovery even if sorbent runs dry
  • Mass spectrometry compatible
  • Copolymer sorbent substrate
  • Mixed mode functionality
  • Not endcapped
  • Ion exchange capacity of 0.6 meq/g
  • Constructed from polypropylene
  • pH range of 0 to 14
Catalog No. 50-819-118 Supplier Waters Corp Supplier No. 186002489
The 1 cc Oasis WAX cartridges contain the Oasis WAX sorbent, a polymeric reversed-phase, weak anion exchange mixed-mode sorbent optimized for fast, simple, and highly selective sample preparation of strong acidic compounds. The Oasis WAX sorbent is water wettable, maintaining its capability for higher retention and excellent recoveries even if the sorbent runs dry, eliminating the need for extraordinary steps to prevent drying out during critical steps prior to sample loading.
  • Polymeric reversed-phase, weak anion exchange mixed-mode sorbent for strong acidic compounds
  • Water wettable to maintain retention and recovery even if sorbent runs dry
  • Mass spectrometry compatible
  • Copolymer sorbent substrate
  • Mixed mode functionality
  • Not endcapped
  • Ion exchange capacity of 0.6 meq/g
  • Constructed from polypropylene
  • pH range of 0 to 14