Learn More
Waters Corp Waters Corporation Vac Cartridge, 3 cc, 60 mg Sorbent, 30 μm, HLB
Catalog No. 50785844 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 3 cc Oasis HLB cartridges offer a universal polymeric reversed-phase sorbent for extracting acidic, basic, and neutral compounds. Water-wettable, it ensures high retention and excellent recoveries even if dried, streamlining sample preparation by eliminating conditioning steps. Intended for automated SPE on the Gilson ASPEC.
  • Universal polymeric reversed-phase sorbent
  • Extracts acidic, basic, and neutral compounds
  • Water-wettable; high retention and recoveries even if dry
  • Eliminates equilibration and conditioning phases
  • Copolymer stable over pH range 0 to 14
  • Automated SPE compatible with Gilson ASPEC
  • Particle size: 30 μm
  • Sorbent weight: 60 mg
  • Barrel size: 3 cc
  • Mass Spec compatible
  • Mode: Reversed Phase
  • Material: Polypropylene
Catalog No. 50-785-844 Supplier Waters Corp Supplier No. WAT058883
The 3 cc Oasis HLB cartridges offer a universal polymeric reversed-phase sorbent for extracting acidic, basic, and neutral compounds. Water-wettable, it ensures high retention and excellent recoveries even if dried, streamlining sample preparation by eliminating conditioning steps. Intended for automated SPE on the Gilson ASPEC.
  • Universal polymeric reversed-phase sorbent
  • Extracts acidic, basic, and neutral compounds
  • Water-wettable; high retention and recoveries even if dry
  • Eliminates equilibration and conditioning phases
  • Copolymer stable over pH range 0 to 14
  • Automated SPE compatible with Gilson ASPEC
  • Particle size: 30 μm
  • Sorbent weight: 60 mg
  • Barrel size: 3 cc
  • Mass Spec compatible
  • Mode: Reversed Phase
  • Material: Polypropylene