Learn More
Aviva Systems Biology Corp OAT Antibody - middle region (ARP48134_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48134_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OAT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human OAT. Alternate names: DKFZp781A11155, HOGA, OKT, GACR, OATASEApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53538