Learn More
Medchemexpress LLC OATD-02 hydrochloride | 2146132-73-0 | 99.9% | 308.61 g/mol | C12H26BClN2O4 | 10 MG
SDP
Catalog No. 5000287377
Encompass_Preferred
OATD-02 hydrochloride is the hydrochloride salt form of an orally active, competitive, reversible, noncovalent dual inhibitor of arginase 1 and arginase 2, intended for preclinical cancer research. It is supplied as a high-purity, water-soluble solid suitable for in vitro and in vivo studies.
  • High purity: 99.9%.
  • White to off-white solid physical form.
  • High aqueous solubility: 250 mg/mL in water (may require ultrasonic).
  • Dual arginase 1 and 2 inhibitory activity for tumor immunology research.
  • Available in small research pack sizes for preclinical use.
Catalog No. 50-002-87377 Supplier Medchemexpress LLC Supplier No. HY155108B10MG
OATD-02 hydrochloride is the hydrochloride salt form of an orally active, competitive, reversible, noncovalent dual inhibitor of arginase 1 and arginase 2, intended for preclinical cancer research. It is supplied as a high-purity, water-soluble solid suitable for in vitro and in vivo studies.
  • High purity: 99.9%.
  • White to off-white solid physical form.
  • High aqueous solubility: 250 mg/mL in water (may require ultrasonic).
  • Dual arginase 1 and 2 inhibitory activity for tumor immunology research.
  • Available in small research pack sizes for preclinical use.