Learn More
MOLTOX Molecular Toxicology Inc Oatmeal Agar, 20 mL/plate, 10 plates/sleeve, Store at Room Temperature
SDP
Catalog No. 50948629
Encompass_Preferred
Oatmeal Agar is used for the cultivation of fungi, particularly macrospores. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-629 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S137
Add to Cart
Add to Cart
Oatmeal Agar is used for the cultivation of fungi, particularly macrospores. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.