Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec OB600 Elevating Workstation
SDP

Catalog No. 22444228 Shop All Mopec Products
Change view
Click to view available options
Height (Metric):
200.1 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric)
22-444-228 200.1 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-444-228 Supplier Mopec Supplier No. OB600

missing translation for 'validMainframeMsg'

One of the most technologically advanced grossing stations available today. Mopec™ OB600 Elevating Workstation combines paramount laboratory work space ventilation and a back-saving elevation feature.

  • All stainless steel construction
  • Includes hot/cold water fixture with wrist blade handles and swing spout faucet and built-In vacuum breaker
  • Recessed stainless steel work surface
  • Removable white polyethylene cutting board 58.4 x 40.6 x 2cm
  • 4 leveling pads
  • Washable ruler with inch/centimeter markings
  • Ruler is located on the inside lip of the workstation
  • Magnetic tool bar holds instruments when not in use
  • Stainless steel c-fold paper towel dispenser
  • 3 energy saving, long life LED lights
  • Backdraft ventilation designed to safely and efficiently remove formaldehyde fumes and hazardous chemical vapors
  • Equipped with a factory installed exhaust duct which can be connected to any in-house ventilation system
  • All plumbing and electrical work accomplished at factory
  • Only trade hookup required

Warranty and Services

1 Year Warranty

Specifications

Product Type Trimming Station
Description Elevating Work Station
Includes One large and one small stainless steel organizer bin
Electrical Requirements GFCI Electrical Receptacle
Frequency 60 Hz
Voltage 115 V
Depth (English) 60 in.
Depth (English) Sink 10 in.
Depth (Metric) 152.4cm
Depth (Metric) Sink 25 cm
Height (English) 79 in.
Width (English) 29 in.
Elevating or Fixed Height Elevating
Height (Metric) 200.1 cm
Length (English) Sink 19 in.
Length (Metric) Sink 48cm
Width (English) Sink 14 in.
Width (Metric) 73.7cm
Width (Metric) Sink 36 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.