Learn More
Medchemexpress LLC HY-N0428 5mg Medchemexpress, Obacunone CAS:751-03-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN04285MG
Encompass_Preferred
Medchemexpress, HY-N0428 5mg Obacunone CAS:751-03-1 Obacunone, isolated from seeds of Marsh White grapefruit, exhibits anti-tumor activity by the induction of apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0269