Learn More
Medchemexpress LLC Obacunone | 751-03-1 | MFCD00075888 | 99.8% | 454.51 g/mol | C26H30O7 | 10 MG
SDP
Catalog No. 5000295777
Encompass_Preferred
Obacunone is a highly oxidized triterpenoid limonoid isolated from citrus plants and supplied as a high-purity research reagent with reported apoptosis-inducing activity and protection against UV-induced oxidative damage in retinal cells.
  • Highly oxidized triterpenoid limonoid
  • Reported to induce apoptosis in cellular models
  • Protects retinal pigment epithelial cells from UV-induced oxidative damage
  • High purity suitable for analytical and research use (99.75%)
  • Chemical formula C26H30O7; molecular weight ~454.5 g/mol
Catalog No. 50-002-95777 Supplier Medchemexpress LLC Supplier No. HYN042810MG
Obacunone is a highly oxidized triterpenoid limonoid isolated from citrus plants and supplied as a high-purity research reagent with reported apoptosis-inducing activity and protection against UV-induced oxidative damage in retinal cells.
  • Highly oxidized triterpenoid limonoid
  • Reported to induce apoptosis in cellular models
  • Protects retinal pigment epithelial cells from UV-induced oxidative damage
  • High purity suitable for analytical and research use (99.75%)
  • Chemical formula C26H30O7; molecular weight ~454.5 g/mol