Learn More
Selleck Chemical LLC Obatoclax Mesylate (GX15-070) S1057-1g
SDP
Catalog No. 503108451
Encompass_Preferred
Obatoclax Mesylate (GX15-070) is an antagonist of Bcl-2 with Ki of 0 22 M in a cell-free assay can assist in overcoming MCL-1 mediated resistance to apoptosis
Catalog No. 50-310-8451 Supplier Selleck Chemical LLC Supplier No. S10571G
Obatoclax Mesylate (GX15-070) is an antagonist of Bcl-2 with Ki of 0 22 M in a cell-free assay can assist in overcoming MCL-1 mediated resistance to apoptosis