Learn More
Selleck Chemical LLC Obatoclax Mesylate (GX15-070) S1057-10mM/1mL
SDP
Catalog No. 503108485
Encompass_Preferred
Obatoclax Mesylate (GX15-070) is an antagonist of Bcl-2 with Ki of 0 22 M in a cell-free assay can assist in overcoming MCL-1 mediated resistance to apoptosis
Catalog No. 50-310-8485 Supplier Selleck Chemical LLC Supplier No. S105710MM/1ML
Obatoclax Mesylate (GX15-070) is an antagonist of Bcl-2 with Ki of 0 22 M in a cell-free assay can assist in overcoming MCL-1 mediated resistance to apoptosis