Learn More
TARGETMOL CHEMICALS INC OBATOCLAX MESYLATE 25MG
SDP
Catalog No. 502576547
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Obatoclax Mesylate (GX15-070) is a Bcl-2 antagonist (Ki 0.22 uM) and can induce apoptosis with up-regulation of Bim induced cytochrome c release and activation of caspase-3. purity: 99%
Catalog No. 50-257-6547 Supplier TARGETMOL CHEMICALS INC Supplier No. T627525MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Obatoclax Mesylate (GX15-070) is a Bcl-2 antagonist (Ki 0.22 uM) and can induce apoptosis with up-regulation of Bim induced cytochrome c release and activation of caspase-3. purity: 99%