Learn More
Medchemexpress LLC Obatoclax mesylate | 803712-79-0 | 99.9% | 413.49 | 1 ML
SDP
Catalog No. 5000344536
Encompass_Preferred
Obatoclax Mesylate is a BH3 mimetic and a pan-BCL-2 family proteins inhibitor that induces autophagy-dependent cell death and targets cyclin D1 for proteasomal degradation. It exhibits anti-cancer and broad-spectrum antiparasitic activity.
  • Inhibits BCL-2 family proteins including BCL-2, BCL-XL, MCL-1, BCL-w, A1, and BCL-b
  • Induces autophagy-dependent cell death
  • Targets cyclin D1 for proteasomal degradation
  • Demonstrates anti-cancer and broad-spectrum antiparasitic activity
  • Reduces cell numbers in human colorectal cancer cell lines (HCT116, HT-29, LoVo)
  • Increases G1-phase cell populations
  • Causes a marked drop in cyclin D1 levels, inhibits GSK3β, and activates p38MAPK
  • Shows potent antitumor activity in xenograft mouse models
Catalog No. 50-003-44536 Supplier Medchemexpress LLC Supplier No. HY1096910MG
Add to Cart
Add to Cart
Obatoclax Mesylate is a BH3 mimetic and a pan-BCL-2 family proteins inhibitor that induces autophagy-dependent cell death and targets cyclin D1 for proteasomal degradation. It exhibits anti-cancer and broad-spectrum antiparasitic activity.
  • Inhibits BCL-2 family proteins including BCL-2, BCL-XL, MCL-1, BCL-w, A1, and BCL-b
  • Induces autophagy-dependent cell death
  • Targets cyclin D1 for proteasomal degradation
  • Demonstrates anti-cancer and broad-spectrum antiparasitic activity
  • Reduces cell numbers in human colorectal cancer cell lines (HCT116, HT-29, LoVo)
  • Increases G1-phase cell populations
  • Causes a marked drop in cyclin D1 levels, inhibits GSK3β, and activates p38MAPK
  • Shows potent antitumor activity in xenograft mouse models