Learn More
Medchemexpress LLC Obatoclax Mesylate | 803712-79-0 | 99.9% | 413.49 | 25 MG
SDP
Catalog No. 5000347671
Encompass_Preferred
Obatoclax Mesylate, also known as GX15-070 Mesylate, is a BH3 mimetic that acts as a pan-BCL-2 family proteins inhibitor with a Ki of 220 nM for BCL-2. It induces autophagy-dependent cell death and targets cyclin D1 for proteasomal degradation. This compound exhibits anti-cancer and broad-spectrum antiparasitic activities.
  • It is a pan-BCL-2 family proteins inhibitor.
  • It induces autophagy-dependent cell death.
  • It targets cyclin D1 for proteasomal degradation.
  • It possesses anti-cancer activity.
  • It demonstrates broad-spectrum antiparasitic activity.
Catalog No. 50-003-47671 Supplier Medchemexpress LLC Supplier No. 25MGHY10969
Obatoclax Mesylate, also known as GX15-070 Mesylate, is a BH3 mimetic that acts as a pan-BCL-2 family proteins inhibitor with a Ki of 220 nM for BCL-2. It induces autophagy-dependent cell death and targets cyclin D1 for proteasomal degradation. This compound exhibits anti-cancer and broad-spectrum antiparasitic activities.
  • It is a pan-BCL-2 family proteins inhibitor.
  • It induces autophagy-dependent cell death.
  • It targets cyclin D1 for proteasomal degradation.
  • It possesses anti-cancer activity.
  • It demonstrates broad-spectrum antiparasitic activity.