Learn More
Selleck Chemical LLC Obatoclax Mesylate (GX15-070) 50mg 803712-79-0
SDP
Supplier:  Selleck Chemical LLC S105750MG
Encompass_Preferred
Obatoclax Mesylate (GX15-070) is an antagonist of Bcl-2 with Ki of 0.22 ?M in a cell-free assay, can assist in overcoming MCL-1 mediated resistance to apoptosis. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-709-6