Learn More
Apexbio Technology LLC Obatoclax mesylate (GX15-070), 803712-79-0, MFCD18074508, 10 mg
SDP
Catalog No. 501014140
Encompass_Preferred
MF: C20H19N3OCH4O3S, MW: 413.5, Solubility: 20.7 mg/mL in DMSO, 3.15 mg/mL in EtOH with ultrasonic and warming,insoluble in H2O. Obatoclax mesylate, also known as GX15-070, is a hydrophobic small molecule that potently inhibits BCL-2 family by binding to the BH3-binding site of BCL-2 and other related BCL-2 family members (including BCL-XL, MCL-1, A1, and BCL-B).
Catalog No. 50-101-4140 Supplier Apexbio Technology LLC Supplier No. A419410
MF: C20H19N3OCH4O3S, MW: 413.5, Solubility: 20.7 mg/mL in DMSO, 3.15 mg/mL in EtOH with ultrasonic and warming,insoluble in H2O. Obatoclax mesylate, also known as GX15-070, is a hydrophobic small molecule that potently inhibits BCL-2 family by binding to the BH3-binding site of BCL-2 and other related BCL-2 family members (including BCL-XL, MCL-1, A1, and BCL-B).