Learn More
Cayman Chemical Obatoclax (mesylate), Purity: ≥98%, CAS: 803712-79-0, 50 mg
SDP
Catalog No. 502954920
Encompass_Preferred
An antagonist of Bcl-2 family members containing four Bcl-2 homology domains, including Bcl-2, Bcl-W, Bcl-xL, and Mcl-1 (Kd ~500 nM); prevents interaction of these pro-survival proteins with Bax or Bak, inducing apoptosis; also induces autophagy in mouse embryo fibroblasts and in HeLa cell
Catalog No. 50-295-4920 Supplier Cayman Chemical Supplier No. 1149950 MG
An antagonist of Bcl-2 family members containing four Bcl-2 homology domains, including Bcl-2, Bcl-W, Bcl-xL, and Mcl-1 (Kd ~500 nM); prevents interaction of these pro-survival proteins with Bax or Bak, inducing apoptosis; also induces autophagy in mouse embryo fibroblasts and in HeLa cell