Learn More
Medchemexpress LLC Obeldesivir | 2647441-36-7 | 99.5% | 361.35 g/mol | C16H19N5O5 | 5 MG
SDP
Catalog No. 5000223058
Encompass_Preferred
Obeldesivir is an orally active ester prodrug of GS-441524 developed for antiviral research. It inhibits replication of SARS-CoV-2 in vitro and is supplied as a high-purity research chemical with defined storage and solubility properties for laboratory use.
  • Orally active antiviral ester prodrug of GS-441524.
  • High purity suitable for research applications.
  • White to off-white solid appearance.
  • Soluble in DMSO at high concentration.
  • Specified storage conditions for solid and solvent solutions.
Catalog No. 50-002-23058 Supplier Medchemexpress LLC Supplier No. HY1459945MG
Obeldesivir is an orally active ester prodrug of GS-441524 developed for antiviral research. It inhibits replication of SARS-CoV-2 in vitro and is supplied as a high-purity research chemical with defined storage and solubility properties for laboratory use.
  • Orally active antiviral ester prodrug of GS-441524.
  • High purity suitable for research applications.
  • White to off-white solid appearance.
  • Soluble in DMSO at high concentration.
  • Specified storage conditions for solid and solvent solutions.