Learn More
MSC Oberon Welding Face Shield and Headgear, Polycarbonate, 1 ea
Catalog No. 5000676562
Encompass
This welding face shield and headgear set provides essential protection for welding applications. It features a polycarbonate window designed for durability and clarity.
  • Polycarbonate window material
  • Green window color
  • Window thickness: 0.0150 in.
  • Window height: 8 in.
  • Window width: 14 in.
  • Lens shade 9-10
  • Ratchet headgear adjustment
  • Meets ANSI Z87.1 standards
Catalog No. 50-006-76562 Supplier MSC Supplier No. 01264589
This welding face shield and headgear set provides essential protection for welding applications. It features a polycarbonate window designed for durability and clarity.
  • Polycarbonate window material
  • Green window color
  • Window thickness: 0.0150 in.
  • Window height: 8 in.
  • Window width: 14 in.
  • Lens shade 9-10
  • Ratchet headgear adjustment
  • Meets ANSI Z87.1 standards