Learn More
CPC Scientific H-Phe-Asn-Ala-Pro-Phe-Asp-Val-Gly-Ile-Lys-Leu-Ser-Gly-Ala-Gln-Tyr-Gln-Gln-His-Gly-Arg-Ala-Leu-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific OBST003A
Encompass
Suppressor of Food Intake and Gastric EmptyingONE-LETTER SEQUENCE: FNAPFDVGIKLSGAQYQQHGRAL-NH2MOLECULAR FORMULA: C114H174N34O31MOLECULAR WEIGHT:2516.85STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [869705-22-6], RESEARCH AREA: GastrointestinalREFERENCES: J.V.Zhang et al., Science, 310, 996 (2005); M. Kojima, H. Hosoda, Y. Date, M. Nakazato, H. Matsuo, and K. Kangawa, Nature, 402, 656 (1999)
Catalog No. 50-210-9968