Learn More
AdipoGen Obestatin human pAb
SDP
Supplier:  AdipoGen AG25A0043C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human obestatin. Cross-reacts with mouse obestatin. Source Rabbit. Applications ELISA. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Obestatin derives like ghrelin from proghrelin, but exhibits opposite physiological effects. Obestatin decreases food intake and body weight in rodents. Moreover, obestatin also counteracts ghrelin’s effect by decelerating gastric emptying and decreasing intestinal contractility. Obestatin binds to orphan G protein-coupled receptor GPR39. Amidation of the C-terminus seems to be important for obestatin’s activity.
Catalog No. 50-256-3268