Learn More
Enzo Life Sciences Obestatin (mouse, rat) (1mg)
SDP
Supplier:  Enzo Life Sciences ALX164005M001
Encompass_Preferred
Food intake modulator. Obestatin derives like ghrelin from proghrelin, but exhibits opposite physiological effects. Obestatin decreases food intake and body weight in rodents. Moreover, obestatin also counteracts ghrelin’s effect by decelerating gastric emptying and decreasing intestinal contractility. Obestatin binds to orphan G protein-coupled receptor GPR39. Amidation of the C-terminus seems to be important for obestatin’s activity. Formula: C114H174N34O31. MW: 2516.8. Purity: ≥97% (HPLC). Formulation: Lyophilized. Peptide content: ≥80%. Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-7852