Learn More
Cayman Chemical ObestatInmouserat EIA KI 96Wel
SDP
Catalog No. 502981803
Encompass_Preferred
Obestatin is an amidated peptide of 23 amino acids with an α-helical secondary conformation. It was first isolated from rat stomach in 2005. Obestatin is a preproghrelin-derived peptide produced by many tissues or organs, including the stomach, pancreas, adipose tissue, skeletal muscle, and the heart. Obestatin was originally identified as an anorexigenic peptide that reduces food intake and body weight by opposing the actions of ghrelin. It has been considered to be an antidiabetic peptide that positively influences glucose and lipid metabolism. It may also play a role in the regulation of blood pressure as its plasma concentration increases in hypertensive patients. Obestatin may also have a role in the regulation of anxiety and memory improvement.
Catalog No. 50-298-1803 Supplier Cayman Chemical Supplier No. 1890396 WELL
Obestatin is an amidated peptide of 23 amino acids with an α-helical secondary conformation. It was first isolated from rat stomach in 2005. Obestatin is a preproghrelin-derived peptide produced by many tissues or organs, including the stomach, pancreas, adipose tissue, skeletal muscle, and the heart. Obestatin was originally identified as an anorexigenic peptide that reduces food intake and body weight by opposing the actions of ghrelin. It has been considered to be an antidiabetic peptide that positively influences glucose and lipid metabolism. It may also play a role in the regulation of blood pressure as its plasma concentration increases in hypertensive patients. Obestatin may also have a role in the regulation of anxiety and memory improvement.