Learn More
Selleck Chemical LLC Obeticholic Acid
SDP
Catalog No. 5000000112
Encompass_Preferred
Obeticholic Acid (INT-747) is a potent and selective farnesoid X receptor (FXR) agonist with EC50 of 99 nM Obeticholic Acid inhibits autophagy Phase 3
Catalog No. 50-000-00112 Supplier Selleck Chemical LLC Supplier No. S76601G
Add to Cart
Add to Cart
Obeticholic Acid (INT-747) is a potent and selective farnesoid X receptor (FXR) agonist with EC50 of 99 nM Obeticholic Acid inhibits autophagy Phase 3