Learn More
Medchemexpress LLC HY-12222 10mg , Obeticholic acid CAS:459789-99-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1222210MG
Encompass_Preferred
HY-12222 10mg Obeticholic acid CAS: 459789-99-2 Obeticholic acid (INT-747) is a potent, selective and orally active FXR agonist with an EC50 of 99 nM. Obeticholic acid has anticholeretic and anti-inflammation effect. Obeticholic acid also induces autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9471