Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Obeticholic acid | 459789-99-2 | 99.9% | 25MG
SDP

Catalog No. 5000231168
Encompass_Preferred

Obeticholic acid | 459789-99-2 | 99.9% | 25MG

Catalog No. 50-002-31168 Supplier Medchemexpress LLC Supplier No. HY12222R25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Obeticholic acid | 459789-99-2 | 99.9% | 25MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.