Learn More
Aviva Systems Biology Corp STN1 Antibody - C-terminal region (ARP62578_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62578_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OBFC1. It was validated on Western Blot. . Alternate names: AAF-44, AAF44, FLJ22559, RP11-541N10.2, RPA-32, STN1, bA541N10.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-287-719