Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC OBI-992 drug-linker | 3039487-10-7 | 96.6% | 2173.46 | C106H162FN9O37 | 10 MG
SDP

Catalog No. 5000295258
Encompass_Preferred

OBI-992 drug-linker is an exatecan-based drug-linker conjugate designed for antibody-drug conjugate (ADC) synthesis. It links the TOP1 inhibitor exatecan to a cleavable linker to enable targeted delivery of the payload; preclinical studies report antitumor activity in mouse and PDX models. Supplied as a solid for research use, the product includes specified purity and storage recommendations for laboratory handling.

  • Composed of exatecan linked to a cleavable linker.
  • Demonstrated antitumor activity in preclinical models.
  • High molecular weight compound suitable for ADC synthesis.
  • Provided as a white to yellow solid.
  • Purity 96.6%.
  • Molecular weight 2173.46.
  • Storage: sealed, away from moisture and light; in solvent -80°C for 6 months, -20°C for 1 month.
  • Supplied in a 10 mg package for research use.

Catalog No. 50-002-95258 Supplier Medchemexpress LLC Supplier No. HY16848910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OBI-992 drug-linker is an exatecan-based drug-linker conjugate designed for antibody-drug conjugate (ADC) synthesis. It links the TOP1 inhibitor exatecan to a cleavable linker to enable targeted delivery of the payload; preclinical studies report antitumor activity in mouse and PDX models. Supplied as a solid for research use, the product includes specified purity and storage recommendations for laboratory handling.

  • Composed of exatecan linked to a cleavable linker.
  • Demonstrated antitumor activity in preclinical models.
  • High molecular weight compound suitable for ADC synthesis.
  • Provided as a white to yellow solid.
  • Purity 96.6%.
  • Molecular weight 2173.46.
  • Storage: sealed, away from moisture and light; in solvent -80°C for 6 months, -20°C for 1 month.
  • Supplied in a 10 mg package for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.