Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Obidoxime chloride, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012644 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2644 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2644 Supplier MilliporeSigma Supelco Supplier No. 7459325MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Obidoxime chloride is a drug, belonging to the group of oximes. It is found to be useful for the treatment of organophosphorus (OP) poisoning, since it has the ability to reverse the binding of organophosphorus compounds to the enzyme acetylcholinesterase (AChE).

TRUSTED_SUSTAINABILITY

Specifications

CAS 114-90-9
Physical Form Neat
Linear Formula C14H16 Cl2N4O3
MDL Number MFCD00038864
Synonym 1,1 ′-(Oxydimethylene)bis(pyridinium-4-carbaldoxime) dichloride; Bis(4-formylpyridiniomethyl) ether dioxime; Toxogonin
Type Analytical Standard
Quantity 25 mg
Formula Weight 359.21
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.