Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Obidoxime dichloride | 114-90-9 | 96.6% | 359.21 | 10 MG
SDP

Catalog No. 5000452193
Encompass_Preferred

Obidoxime dichloride is a non-full spectrum oxime agent used as an antidote for organophosphate nerve agent poisoning. It reactivates sarin-inhibited acetylcholinesterase (AChE) and reduces the acute toxicity of sarin. It is for research use only and not sold to patients. In vivo, obidoxime dichloride (intramuscular injection; 10 mg/kg; after 60 mins) exhibits an LD50 of 57.7 mg/kg after administration of sarin, and it reduces the acute toxicity of sarin.

  • Used as an antidote for organophosphate nerve agent poisoning
  • Reactivates sarin-inhibited acetylcholinesterase (AChE)
  • Reduces the acute toxicity of sarin
  • Off-white to light yellow solid appearance
  • Soluble in water and DMSO
  • Ships at room temperature in continental US
  • Storage conditions: -20°C, protect from light, stored under nitrogen

Catalog No. 50-004-52193 Supplier Medchemexpress LLC Supplier No. HYW01110810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Obidoxime dichloride is a non-full spectrum oxime agent used as an antidote for organophosphate nerve agent poisoning. It reactivates sarin-inhibited acetylcholinesterase (AChE) and reduces the acute toxicity of sarin. It is for research use only and not sold to patients. In vivo, obidoxime dichloride (intramuscular injection; 10 mg/kg; after 60 mins) exhibits an LD50 of 57.7 mg/kg after administration of sarin, and it reduces the acute toxicity of sarin.

  • Used as an antidote for organophosphate nerve agent poisoning
  • Reactivates sarin-inhibited acetylcholinesterase (AChE)
  • Reduces the acute toxicity of sarin
  • Off-white to light yellow solid appearance
  • Soluble in water and DMSO
  • Ships at room temperature in continental US
  • Storage conditions: -20°C, protect from light, stored under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.