Learn More
Cayman Chemical Obidoxime (chloride), Purity: ≥95%, CAS: 114-90-9, 1 g
SDP
Catalog No. 502913803
Encompass_Preferred
An AChE modulator; induces inhibition and activation of bovine erythrocyte AChE in the presence and absence of ACh, respectively, at 3 mM; induces reactivation of tabun-inhibited AChE in rat blood and diaphragm and reduces tabun-induced lethality in mice at 10.5 mg/kg; intranasal administration reduces seizure duration and severity and prevents mortality in a rat model of paraoxon-induced organophosphate poisoning
Catalog No. 50-291-3803 Supplier Cayman Chemical Supplier No. 365871 G
An AChE modulator; induces inhibition and activation of bovine erythrocyte AChE in the presence and absence of ACh, respectively, at 3 mM; induces reactivation of tabun-inhibited AChE in rat blood and diaphragm and reduces tabun-induced lethality in mice at 10.5 mg/kg; intranasal administration reduces seizure duration and severity and prevents mortality in a rat model of paraoxon-induced organophosphate poisoning