Learn More
Leica Biosystems HC PL FLUOTAR 20x/0.55 Microscope Objective (20x, NA 0.55)
Catalog No. NC3366050
Encompass_Preferred
HC PL FLUOTAR 20x/0.55 is a fluorescence-optimized microscope objective that provides 20x magnification with a 0.55 numerical aperture. It delivers flat-field correction and chromatic control suitable for fluorescence and transmitted-light imaging in routine and research microscopy workflows.
  • 20x magnification with 0.55 numerical aperture.
  • Fluorescence-optimized optical design for high-contrast imaging.
  • Plan correction for a flat, well-corrected field of view.
  • Appropriate for routine and advanced light-microscopy applications.
  • Compatible with standard microscope objective turrets and parfocal systems.
Catalog No. NC3366050 Supplier Leica Biosystems Supplier No. 11506519
HC PL FLUOTAR 20x/0.55 is a fluorescence-optimized microscope objective that provides 20x magnification with a 0.55 numerical aperture. It delivers flat-field correction and chromatic control suitable for fluorescence and transmitted-light imaging in routine and research microscopy workflows.
  • 20x magnification with 0.55 numerical aperture.
  • Fluorescence-optimized optical design for high-contrast imaging.
  • Plan correction for a flat, well-corrected field of view.
  • Appropriate for routine and advanced light-microscopy applications.
  • Compatible with standard microscope objective turrets and parfocal systems.