Learn More
Leica Biosystems Leica Microsystems HC PL Fluotar 40x/0.80 Dry Fluorescence Objective
Catalog No. NC3366053
Encompass_Preferred
The HC PL Fluotar 40x/0.80 dry fluorescence objective delivers high transmission and chromatic correction for fluorescence imaging, providing strong photon collection and high-resolution contrast for fixed and stained specimens.
  • 40x magnification with a 0.80 numerical aperture.
  • Dry objective designed for fluorescence microscopy applications.
  • PL FLUOTAR optical correction for improved chromatic performance.
  • High transmission for enhanced photon collection in low-light imaging.
  • Short free working distance of 0.4 mm for high-resolution imaging.
  • Standard M25 objective thread for compatibility with Leica and compatible microscope systems.
Catalog No. NC3366053 Supplier Leica Biosystems Supplier No. 11506382
The HC PL Fluotar 40x/0.80 dry fluorescence objective delivers high transmission and chromatic correction for fluorescence imaging, providing strong photon collection and high-resolution contrast for fixed and stained specimens.
  • 40x magnification with a 0.80 numerical aperture.
  • Dry objective designed for fluorescence microscopy applications.
  • PL FLUOTAR optical correction for improved chromatic performance.
  • High transmission for enhanced photon collection in low-light imaging.
  • Short free working distance of 0.4 mm for high-resolution imaging.
  • Standard M25 objective thread for compatibility with Leica and compatible microscope systems.