Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Motic™ Objective 2.0X ESD for SMZ161/171

Catalog No. 01550013 Shop All Motic Group Products
Change view
Click to view available options
Magnification Power:
2X
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
01-550-013 2X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-550-013 Supplier Motic™ Supplier No. 1101001703681
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Doubles the standard magnification. Motic™ Objective Lens 2.0X ESD is used with SMZ161/171 StereoZoom Microscopes

  • Doubles the standard magnification
  • WD=38.6mm
  • Includes Electrostatic Discharge feature
  • For use with SMZ161/171 StereoZoom microscope

Order Info

dimProductType

Specifications

Type Lens
Focus Distance 38.6 mm WD
For Use With (Equipment) SMZ161/171
Includes Electrostatic Discharge feature
Magnification Power 2X
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.