Learn More
Molecular Devices Molecular Devices SpectraCuvette Adapter, 18 mm, SpectraMax compatible
Catalog No. 5000014662
Encompass_Preferred
The Molecular Devices SpectraCuvette Adapter (part number 5301193) adapts standard cuvettes for measurement in compatible microplate readers by positioning cuvettes at a defined plate height for accurate optical alignment and absorbance readings.
  • Provides 18 mm plate height for proper optical alignment.
  • Holds cuvettes securely for absorbance measurements.
  • Designed to maintain plate height below instrument maximums to prevent damage.
  • Uses plate-height marking to ensure correct adapter selection.
Catalog No. 50-000-14662 Supplier Molecular Devices Supplier No. 5301193
The Molecular Devices SpectraCuvette Adapter (part number 5301193) adapts standard cuvettes for measurement in compatible microplate readers by positioning cuvettes at a defined plate height for accurate optical alignment and absorbance readings.
  • Provides 18 mm plate height for proper optical alignment.
  • Holds cuvettes securely for absorbance measurements.
  • Designed to maintain plate height below instrument maximums to prevent damage.
  • Uses plate-height marking to ensure correct adapter selection.