Learn More
Molecular Devices Nikon Microscope Objective, 40X, 0.60 NA, ELWD
Catalog No. 5000014689
Encompass_Preferred
The Nikon CFI S Plan Fluor ELWD 40XC is a 40X fluorescence objective designed for extra-long working distance imaging in air. It is suitable for brightfield, fluorescence, and apodized phase contrast applications, and includes cover-glass correction for variable sample thickness.
  • 40X magnification with 0.60 numerical aperture.
  • Extra-long working distance, 3.6-2.8 mm, for imaging thick samples or through plates.
  • Cover-glass correction range 0-2 mm to accommodate varying sample preparations.
  • Designed for air (dry) use, compatible with standard infinity-corrected microscope systems.
Catalog No. 50-000-14689 Supplier Molecular Devices Supplier No. 65000109
The Nikon CFI S Plan Fluor ELWD 40XC is a 40X fluorescence objective designed for extra-long working distance imaging in air. It is suitable for brightfield, fluorescence, and apodized phase contrast applications, and includes cover-glass correction for variable sample thickness.
  • 40X magnification with 0.60 numerical aperture.
  • Extra-long working distance, 3.6-2.8 mm, for imaging thick samples or through plates.
  • Cover-glass correction range 0-2 mm to accommodate varying sample preparations.
  • Designed for air (dry) use, compatible with standard infinity-corrected microscope systems.