Learn More
Molecular Devices Objective 4x Plan Apo Lambda D, 0.2 NA
Catalog No. 5000014705
Encompass_Preferred
Objective 4x Plan Apo Lambda D, 0.2 NA.
Catalog No. 50-000-14705 Supplier Molecular Devices Supplier No. 5301119
Add to Cart
Add to Cart
Objective 4x Plan Apo Lambda D, 0.2 NA.