Learn More
Molecular Devices Nikon Objective, CFI S Plan Fluor ELWD, 60X, 0.7 NA
Catalog No. 5000014692
Encompass_Preferred
60X extra-long working distance objective with 0.70 numerical aperture, optimized for fluorescence and brightfield imaging. The correction collar accommodates variable cover glass thickness and the extended working distance suits thick samples and culture vessels.
  • 60x magnification with 0.70 numerical aperture.
  • Extra-long working distance for imaging thick samples.
  • Correction collar for 0.1-1.3 mm cover glass thickness.
  • Air immersion, spring-loaded objective design.
  • Suitable for fluorescence and brightfield applications.
  • Compatible with high-content imaging and live-cell workflows.
Catalog No. 50-000-14692 Supplier Molecular Devices Supplier No. 65000110
Add to Cart
Add to Cart
Request a Quote
60X extra-long working distance objective with 0.70 numerical aperture, optimized for fluorescence and brightfield imaging. The correction collar accommodates variable cover glass thickness and the extended working distance suits thick samples and culture vessels.
  • 60x magnification with 0.70 numerical aperture.
  • Extra-long working distance for imaging thick samples.
  • Correction collar for 0.1-1.3 mm cover glass thickness.
  • Air immersion, spring-loaded objective design.
  • Suitable for fluorescence and brightfield applications.
  • Compatible with high-content imaging and live-cell workflows.