Learn More
Molecular Devices Molecular Devices Microscope Objective, 60X, Plan Fluor, 0.85 NA
Catalog No. 5000014713
Encompass_Preferred
This high-magnification Plan Fluor objective is designed for fluorescence imaging on automated high-content systems, providing a balance of resolution and light-gathering for detailed cellular imaging.
  • 60x magnification for high-resolution imaging.
  • 0.85 numerical aperture for increased light-gathering and resolution.
  • Air immersion, no immersion medium required.
  • Designed for thin coverslip specimens (~0.17 mm).
  • Optimized for fluorescence applications and automated objective turrets.
Catalog No. 50-000-14713 Supplier Molecular Devices Supplier No. 163000414
This high-magnification Plan Fluor objective is designed for fluorescence imaging on automated high-content systems, providing a balance of resolution and light-gathering for detailed cellular imaging.
  • 60x magnification for high-resolution imaging.
  • 0.85 numerical aperture for increased light-gathering and resolution.
  • Air immersion, no immersion medium required.
  • Designed for thin coverslip specimens (~0.17 mm).
  • Optimized for fluorescence applications and automated objective turrets.