Learn More
Molecular Devices Molecular Devices Filter Lens, 405 nm, Optical, Microplate Reader Compatible
Catalog No. 5000014727
Encompass_Preferred
Molecular Devices 405 nm filter lens is an optical bandpass filter designed for filter-based microplate readers. It transmits light centered at 405 nm for selective detection in absorbance or fluorescence assays and serves as a replaceable filter accessory compatible with common filter-based readers such as EMax, VMax, UVMax, and ThermoMax models.
  • Center wavelength 405 nm.
  • Optical bandpass filter lens for filter-based detection.
  • Replaceable accessory for microplate readers.
  • Suitable for assays requiring detection near 405 nm.
Catalog No. 50-000-14727 Supplier Molecular Devices Supplier No. 65000006
Molecular Devices 405 nm filter lens is an optical bandpass filter designed for filter-based microplate readers. It transmits light centered at 405 nm for selective detection in absorbance or fluorescence assays and serves as a replaceable filter accessory compatible with common filter-based readers such as EMax, VMax, UVMax, and ThermoMax models.
  • Center wavelength 405 nm.
  • Optical bandpass filter lens for filter-based detection.
  • Replaceable accessory for microplate readers.
  • Suitable for assays requiring detection near 405 nm.