Learn More
Molecular Devices Molecular Devices Absorbance Filter, 260 nm, 5.5 nm Bandwidth
Catalog No. 5000014710
Encompass_Preferred
The Molecular Devices 260 nm absorbance filter (5.5 nm bandwidth) is an optical bandpass accessory designed for use with compatible FilterMax series multimode microplate readers. It provides a narrow UV transmission centered at 260 nm for precise absorbance measurements of nucleic acids and other UV-absorbing analytes.
  • Provides narrow bandpass transmission centered at 260 nm.
  • 5.5 nm bandwidth for selective UV absorbance measurements.
  • Suitable for nucleic acid quantification and other UV-absorbing samples.
  • Designed for easy installation in modular filter holders.
  • Improves measurement specificity by reducing out-of-band light.
Catalog No. 50-000-14710 Supplier Molecular Devices Supplier No. 65900001
The Molecular Devices 260 nm absorbance filter (5.5 nm bandwidth) is an optical bandpass accessory designed for use with compatible FilterMax series multimode microplate readers. It provides a narrow UV transmission centered at 260 nm for precise absorbance measurements of nucleic acids and other UV-absorbing analytes.
  • Provides narrow bandpass transmission centered at 260 nm.
  • 5.5 nm bandwidth for selective UV absorbance measurements.
  • Suitable for nucleic acid quantification and other UV-absorbing samples.
  • Designed for easy installation in modular filter holders.
  • Improves measurement specificity by reducing out-of-band light.