Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Motic™ Objectives, Stereo for SMZ-161/171

Catalog No. 01550016 Shop All Motic Group Products
Change view
Click to view available options
Magnification Power:
0.63X
0.75X
1.5X
Description:
WD: 128.6mm
WD: 142.7mm
WD: 56.3mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power Description
01-550-016 0.63X WD: 142.7mm
01-550-015 1.5X WD: 56.3mm
01-550-011 0.75X WD: 128.6mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-550-016 Supplier Motic™ Supplier No. 1101001703661
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable stereo objectives for SMZ-161 and SMZ-171 Microscopes. Motic™ Objectives, Stereo for SMZ-161/171 are precision-crafted for superior quality and durability.

  • Suitable for SMZ-161 and SMZ-171 Microscopes.

Specifications

Type Stereo
Description WD: 142.7mm
For Use With (Equipment) SMZ-161/171 Microscopes
Magnification Power 0.63X
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.