Learn More
Medchemexpress LLC Oblimersen sodium | 98.09% | 5684.61 (free acid) | 1 MG
SDP
Catalog No. 5000437114
Encompass_Preferred
Oblimersen sodium functions as a BCL-2 inhibitor by targeting BCL-2 RNA. It specifically binds to the first six codons of the bcl-2 mRNA sequence, leading to the degradation of bcl-2 mRNA and inducing apoptosis by down-regulating the expression of Bcl-2. Oblimersen sodium can be used for cancer research.
  • Functions as a BCL-2 inhibitor
  • Targets BCL-2 RNA
  • Binds to the first six codons of bcl-2 mRNA sequence
  • Leads to degradation of bcl-2 mRNA
  • Induces apoptosis by down-regulating Bcl-2 expression
  • Suitable for cancer research
Catalog No. 50-004-37114 Supplier Medchemexpress LLC Supplier No. HY118874A1MG
Oblimersen sodium functions as a BCL-2 inhibitor by targeting BCL-2 RNA. It specifically binds to the first six codons of the bcl-2 mRNA sequence, leading to the degradation of bcl-2 mRNA and inducing apoptosis by down-regulating the expression of Bcl-2. Oblimersen sodium can be used for cancer research.
  • Functions as a BCL-2 inhibitor
  • Targets BCL-2 RNA
  • Binds to the first six codons of bcl-2 mRNA sequence
  • Leads to degradation of bcl-2 mRNA
  • Induces apoptosis by down-regulating Bcl-2 expression
  • Suitable for cancer research