Learn More
AdipoGen Obscurolide A1
SDP
Supplier:  AdipoGen BVT0287M001
Encompass_Preferred
Chemical. CAS 144397-99-9. Formula C15H17NO5. MW 291.3. Isolated from Streptomyces viridochromogenes. Weak phosphodiesterase inhibitor.
Catalog No. 50-256-2013