Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Obtucarbamate A | 6935-99-5 | 1 ML
SDP

Catalog No. 5000421700
Encompass_Preferred

Obtucarbamate A, isolated from *Disporum cantoniense*, is an off-white to gray solid demonstrating antitussive activity. With a purity of 99.59%, it exhibits antineuroinflammatory activity in mouse BV2 cells, inhibiting LPS-induced NO production with an IC50 of 10.57 μM. This product is intended for research use only and is not for sale to patients.

  • Isolated from *Disporum cantoniense*
  • Possesses antitussive activity
  • Purity of 99.59%
  • Molecular weight: 238.24
  • Chemical formula: C11H14N2O4
  • Appears as an off-white to gray solid
  • Exhibits antineuroinflammatory activity in mouse BV2 cells
  • Inhibits LPS-induced NO production with an IC50 of 10.57 μM
  • Intended for research use only

Catalog No. 50-004-21700 Supplier Medchemexpress LLC Supplier No. HYN316310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Obtucarbamate A, isolated from *Disporum cantoniense*, is an off-white to gray solid demonstrating antitussive activity. With a purity of 99.59%, it exhibits antineuroinflammatory activity in mouse BV2 cells, inhibiting LPS-induced NO production with an IC50 of 10.57 μM. This product is intended for research use only and is not for sale to patients.

  • Isolated from *Disporum cantoniense*
  • Possesses antitussive activity
  • Purity of 99.59%
  • Molecular weight: 238.24
  • Chemical formula: C11H14N2O4
  • Appears as an off-white to gray solid
  • Exhibits antineuroinflammatory activity in mouse BV2 cells
  • Inhibits LPS-induced NO production with an IC50 of 10.57 μM
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.