Learn More
Medchemexpress LLC Obtusifolin-2-O-glucoside | 120163-18-0 | 446.40 | 1 MG
SDP
Catalog No. 5000415610
Encompass_Preferred
Obtusifolin-2-O-glucoside, identified as compound 7, is a Tyrosinase inhibitor with an IC50 of 9.2 μM. It can be isolated from cassia seed and has been shown to inhibit melanin production in α-MSH-treated B16F10 melanoma cells.
  • Tyrosinase inhibitor (IC50=9.2 μM)
  • Can be isolated from cassia seed
  • Inhibits melanin production in α-MSH-treated B16F10 melanoma cells
  • Purity: 95.30%
Catalog No. 50-004-15610 Supplier Medchemexpress LLC Supplier No. HYN110631MG
Obtusifolin-2-O-glucoside, identified as compound 7, is a Tyrosinase inhibitor with an IC50 of 9.2 μM. It can be isolated from cassia seed and has been shown to inhibit melanin production in α-MSH-treated B16F10 melanoma cells.
  • Tyrosinase inhibitor (IC50=9.2 μM)
  • Can be isolated from cassia seed
  • Inhibits melanin production in α-MSH-treated B16F10 melanoma cells
  • Purity: 95.30%